  • Search Results
  • In vitro and in vivo antimutagenic effects of DIG, a herbal preparation of Berberis vulgaris, Taraxacum officinale and Arctium lappa, against mitomycin C.
In vitro and in vivo antimutagenic effects of DIG, a herbal preparation of Berberis vulgaris, Taraxacum officinale and Arctium lappa, against mitomycin C.
Journal of natural medicines (2015-02-11)
C Di Giorgio, L Boyer, M De Meo, C Laurant, R Elias, E Ollivier
ABSTRACT
DIG, a liquid herbal preparation made from a mixture of diluted mother tinctures of Berberis vulgaris, Taraxacum officinale and Arctium lappa, was assessed for its antimutagenic properties against mitomycin C. The micronucleus assay on Chinese hamster ovary (CHO)-K1 cells was used to evaluate the in vitro anticlastogenic activity of DIG compared to those of separately diluted mother tinctures. The micronucleus assay was performed on mouse erythrocytes and the comet assay was performed on mouse liver, kidney, lung, brain and testicles to assess the protective effects of DIG (0.2 and 2 % at libitum) against an intraperitoneal injection of mitomycin C (1 mg Kg(-1)) in mice. DIG exerted a powerful anticlastogenic activity, under both pretreatment and simultaneous treatment conditions as assessed by the micronucleus assay in CHO-K1 cells. Its protective activity was greater than that observed for each mother tincture. DIG reduced micronuclei levels in mouse erythrocytes and suppressed >80 % of DNA strand breaks in the liver, kidney, lung, brain and testicles of mice exposed to mitomycin C.
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, configured for PerkinElmer 8900, configured for Polygen, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥970 μg/mg (USP XXIV)
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), potency: ≥970 μg per mg (USP XXIV), γ-irradiated, suitable for cell culture
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)