  • Search Results
  • Design, synthesis, and delivery studies of organotin(IV) based HCV inhibitor.
Design, synthesis, and delivery studies of organotin(IV) based HCV inhibitor.
Infectious disorders drug targets (2015-09-04)
Farooq A Shah, Kaneez Fatima, Saqib Ali, Ishtiaq Qadri
ABSTRACT
Tributylstannic[3-(3,5 -dimethylphenylamido)propionate] is synthesized and characterized by elemental analysis, FT-IR, multinuclear NMR ((1)H, (13)C and (119)Sn) and mass spectrometry. The organic anion was found to act as monodentate O-bound ligand in solution. The compound was screened for the anti-HCV potency by the Gaussia luciferase Assay using infected Huh 7.5 cells (human hepatocellular cell) and is found active against HCV with logIC50 1.2nM in the cell-based assay. Cationic surfactant cetyl N,N,N-trimethylammoniumbromide (CTAB) was used to study the interactions of the organotin(IV) complex with positively charged micelles of the surfactant acting as a model cell membrane. The thermodynamics parameters of complex- CTAB interaction concluded that the complex is located in the palisade layer of CTAB micelles. The increase in absorbance of visible spectra of the compound confirmed its solubilization into micelles. The two carbonyl oxygen's were found to be binding sites of the complex with CTAB.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Maleic anhydride, 99%
Sigma-Aldrich
3,5-Dimethylaniline, 98%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Maleic anhydride, powder, 95%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%