  • Search Results
  • Suppression of multidrug resistance by rosiglitazone treatment in human ovarian cancer cells through downregulation of FZD1 and MDR1 genes.
Suppression of multidrug resistance by rosiglitazone treatment in human ovarian cancer cells through downregulation of FZD1 and MDR1 genes.
Anti-cancer drugs (2015-06-09)
Hui Zhang, Xuanxuan Jing, Xiaojuan Wu, Jing Hu, Xiaofang Zhang, Xiao Wang, Peng Su, Weiwei Li, Gengyin Zhou
ABSTRACT
Multidrug resistance (MDR) is a major obstacle in the successful treatment of ovarian cancer. One of the most common causes of MDR is overexpression of P-glycoprotein (P-gp), encoded by the MDR1 gene. The MDR1 gene is a direct target of the Wnt/β-catenin signaling pathway, which plays an important role in ovarian cancer. Peroxisome proliferator-activated receptor γ (PPARγ) ligands have been found to protect against development of cancer through the Wnt/β-catenin pathway. To investigate the effect of PPARγ ligands on MDR1/P-gp expression, we treated a MDR ovarian cancer cell subline, A2780/Taxol, with different concentrations of rosiglitazone (Rosi), a member of the synthetic PPARγ ligands. Rosi downregulated FZD1 and MDR1/P-gp expression in a concentration-dependent manner. In addition, nuclear β-catenin levels and its transcriptional activity decreased significantly. In conclusion, Rosi may reverse MDR of ovarian cancer cells by downregulating the Wnt/β-catenin pathway with the suppression of FZD1.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥97%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
GW9662, >98% (HPLC)
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
2-Chloro-5-nitrobenzanilide, 97%