  • Search Results
  • Comparative characterization of nucleotides, nucleosides and nucleobases in Abelmoschus manihot roots, stems, leaves and flowers during different growth periods by UPLC-TQ-MS/MS.
Comparative characterization of nucleotides, nucleosides and nucleobases in Abelmoschus manihot roots, stems, leaves and flowers during different growth periods by UPLC-TQ-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-11-10)
Le-Yue Du, Da-Wei Qian, Shu Jiang, Er-Xin Shang, Jian-Ming Guo, Pei Liu, Shu-Lan Su, Jin-Ao Duan, Min Zhao
ABSTRACT
Nucleotides, nucleosides and nucleobases have been proven as important bioactive compounds related to many physiological processes. Abelmoschus manihot (L.) Medicus from the family of Malvaceae is an annual herbal plant of folk medicine widely distributed in Oceania and Asia. However, up to now, no detailed information could be available for the types and contents of nucleotides, nucleosides and nucleobases contained in A. manihot roots, stems, leaves as well as the flowers. In the present study, an UPLC-TQ-MS/MS method was established for detection of the twelve nucleotides, nucleosides and nucleobases. The validated method was successfully applied to identify the 12 analytes in different parts of A. manihot harvested at ten growth periods. 2'-deoxyinosine was not detected in all of the A. manihot samples. The data demonstrated that the distribution and concentration of the 12 compounds in A. manihot four parts were arranged in a decreasing order as leaf>flower>stem>root. Based on the results, the leaves and flowers of A. manihot could be developed as health products possessed nutraceutical and bioactive properties in the future. This method might also be utilized for the quality control of the A. manihot leaves and other herbal medicines being rich in nucleotides, nucleosides and nulecobases.
Sigma-Aldrich
Cytidine 5′-monophosphate, Sigma Grade, ≥99% (HPLC), synthetic, powder
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Thymine, suitable for cell culture, BioReagent
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymine, ≥99%
Sigma-Aldrich
2′-Deoxyinosine, ≥98%
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, configured for PerkinElmer 8900, configured for Polygen, for DNA synthesis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)