  • Search Results
  • The effect of Curcuma longa extracted (curcumin) on the quality of cryopreserved boar semen.
The effect of Curcuma longa extracted (curcumin) on the quality of cryopreserved boar semen.
Animal science journal = Nihon chikusan Gakkaiho (2015-06-03)
Panida Chanapiwat, Kampon Kaeoket
ABSTRACT
The aim of this study was to determine the optimal concentration of curcumin needed for cryopreservation of boar semen. Semen samples (n = 9) were collected from nine Duroc boars which having proven fertility were used for routine artificial insemination. Semen samples were collected and divided into six groups (groups A-F) according to various concentrations of curcumin in freezing extender (i.e. 0, 0.125, 0.25, 0.50, 0.75 and 1.0 mmol/L, respectively). The semen was frozen by traditional liquid nitrogen vapor method and stored at -196°C in the liquid nitrogen tank. After storage, frozen semen samples were thawed at 50°C for 12 s and evaluated for progressive motility, viability and acrosome integrity. The present results indicated that the addition of curcumin at 0.25 (group C) or 0.50 mmol/L curcumin (group D) yielded the higher percentage of progressive motility (33.3 and 36.1%, respectively) (P < 0.001). A significantly higher percentage of acrosome integrity was found in groups B (29.7%), C (31.1%) and D (30.2%) than in the other groups (P < 0.01). However, there was no significant difference in percentage of viability among groups. In conclusion, addition to the freezing extender of curcumin during cryopreservation at a concentration of 0.25 or 0.50 mmol/L is the optimal concentration of curcumin for improving the quality (i.e. increased progressive motility and acrosome integrity) of cryopreserved boar semen.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)