  • Search Results
  • Colocalization and Sequential Enzyme Activity in Aqueous Biphasic Systems: Experiments and Modeling.
Colocalization and Sequential Enzyme Activity in Aqueous Biphasic Systems: Experiments and Modeling.
Biophysical journal (2015-11-21)
Bradley W Davis, William M Aumiller, Negar Hashemian, Songon An, Antonios Armaou, Christine D Keating
ABSTRACT
Subcellular compartmentalization of biomolecules and their reactions is common in biology and provides a general strategy for improving and/or controlling kinetics in metabolic pathways that contain multiple sequential enzymes. Enzymes can be colocalized in multiprotein complexes, on scaffolds or inside subcellular organelles. Liquid organelles formed by intracellular phase coexistence could provide an additional means of sequential enzyme colocalization. Here we use experiment and computation to explore the kinetic consequences of sequential enzyme compartmentalization into model liquid organelles in a crowded polymer solution. Two proteins of the de novo purine biosynthesis pathway, ASL (adenylosuccinate lyase, Step 8) and ATIC (5-aminoimidazole-4-carboxamide ribonucleotide transformylase/inosine monophosphate cyclohydrolase, Steps 9 and 10), were studied in a polyethylene glycol/dextran aqueous two-phase system. Dextran-rich phase droplets served as model liquid compartments for enzyme colocalization. In this system, which lacks any specific binding interactions between the phase-forming polymers and the enzymes, we did not observe significant rate enhancements from colocalization for the overall reaction under our experimental conditions. The experimental results were used to adapt a mathematical model to quantitatively describe the kinetics. The mathematical model was then used to explore additional, experimentally inaccessible conditions to predict when increased local concentrations of enzymes and substrates can (or cannot) be expected to yield increased rates of product formation. Our findings indicate that colocalization within these simplified model liquid organelles can lead to enhanced metabolic rates under some conditions, but that very strong partitioning into the phase that serves as the compartment is necessary. In vivo, this could be provided by specific binding affinities between components of the liquid compartment and the molecules to be localized within it.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Fumaric acid, FCC, FG
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ~99%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, meets USP testing specifications, powder
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free