  • Search Results
  • Genomic evidence reveals numerous Salmonella enterica serovar Newport reintroduction events in Suwannee watershed irrigation ponds.
Genomic evidence reveals numerous Salmonella enterica serovar Newport reintroduction events in Suwannee watershed irrigation ponds.
Applied and environmental microbiology (2015-09-20)
Baoguang Li, Scott A Jackson, Jayanthi Gangiredla, Weimin Wang, Huanli Liu, Ben D Tall, Junia Jean-Gilles Beaubrun, Michele Jay-Russell, George Vellidis, Christopher A Elkins
ABSTRACT
Our previous work indicated a predominance (56.8%) of Salmonella enterica serovar Newport among isolates recovered from irrigation ponds used in produce farms over a 2-year period (B. Li et al., Appl Environ Microbiol 80:6355-6365, http://dx.doi.org/10.1128/AEM.02063-14). This observation provided a valuable set of metrics to explore an underaddressed issue of environmental survival of Salmonella by DNA microarray. Microarray analysis correctly identified all the isolates (n = 53) and differentiated the S. Newport isolates into two phylogenetic lineages (S. Newport II and S. Newport III). Serovar distribution analysis showed no instances where the same serovar was recovered from a pond for more than a month. Furthermore, during the study, numerous isolates with an indistinguishable genotype were recovered from different ponds as far as 180 km apart for time intervals as long as 2 years. Although isolates within either lineage were phylogenetically related as determined by microarray analysis, subtle genotypic differences were detected within the lineages, suggesting that isolates in either lineage could have come from several unique hosts. For example, strains in four different subgroups (A, B, C, and D) possessed an indistinguishable genotype within their subgroups as measured by gene differences, suggesting that strains in each subgroup shared a common host. Based on this comparative genomic evidence and the spatial and temporal factors, we speculated that the presence of Salmonella in the ponds was likely due to numerous punctuated reintroduction events associated with several different but common hosts in the environment. These findings may have implications for the development of strategies for efficient and safe irrigation to minimize the risk of Salmonella outbreaks associated with fresh produce.
Sigma-Aldrich
CPNQ, ≥98% (HPLC), solid
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%