  • Search Results
  • Metal and Metal Carbide Nanoparticle Synthesis Using Electrical Explosion of Wires Coupled with Epoxide Polymerization Capping.
Metal and Metal Carbide Nanoparticle Synthesis Using Electrical Explosion of Wires Coupled with Epoxide Polymerization Capping.
Inorganic chemistry (2015-05-27)
Elseddik M Abdelkader, Paul A Jelliss, Steven W Buckner
ABSTRACT
In this study, metal-containing nanoparticles (NPs) were produced using electrical explosion of wires (EEW) in organic solvents. The explosion chamber was constructed from Teflon to withstand the shockwave, allow growth and reaction of the incipient NPs in various organic solvents containing dissolved ligands, and allow a constant flow of argon to maintain an inert environment. A survey of different transition d-block metals was conducted with metals from groups 4-8, affording metal carbide NPs, while metals from groups 9-12 gave elemental metallic NPs. Tungsten carbide phase WC1-x, which has not been previously isolated as a single-phase material, was exclusively formed during EEW. We used polymerization initiation by electron-rich metallic nanoparticles (PIERMEN) as a capping technique for the nascent NPs with an alkyl epoxide employed as the monomers. Transmission electron microscopy showed spherical particles with the metallic core embedded in a polymer matrix with predominantly smaller particles (<50 nm), but also a broad size distribution with some larger particles (>100 nm). Powder X-ray diffraction (PXRD) was used to confirm the identity of the metallic NPs. The capping agents were characterized using ATR-FTIR spectroscopy. No evidence is observed for the formation of crystalline oxides during EEW for any metals used. Differential scanning calorimetry/thermal gravimetric analysis was used to study the NP's behavior upon heating under an air flow up to 800 °C with the product oxides characterized by PXRD. The bifurcation between metal-carbide NPs and metal NPs correlates with the enthalpy of formation of the product carbides. We observed PIERMEN capping of elemental metal NPs only when the metal has negative standard electrode potentials (relative to a bis(biphenyl) chromium(I)/(0) reference electrode).
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Diethylenetriamine, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, rod, diam. 6.2 mm, 99.7% trace metals basis
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
Sigma-Aldrich
Acetonitrile
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%