  • Search Results
  • Comparative proteomic analysis using 2DE-LC-MS/MS reveals the mechanism of Fuzhuan brick tea extract against hepatic fat accumulation in rats with nonalcoholic fatty liver disease.
Comparative proteomic analysis using 2DE-LC-MS/MS reveals the mechanism of Fuzhuan brick tea extract against hepatic fat accumulation in rats with nonalcoholic fatty liver disease.
Electrophoresis (2015-06-04)
Zhonghua Liu, Yong Lin, Sheng Zhang, Die Wang, Qionglin Liang, Guoan Luo
ABSTRACT
Fuzhuan brick tea has received increasing attention in recent years owing to its benefits for nonalcoholic fatty liver disease (NAFLD) and associated metabolic syndrome. For exploring the ameliorative mechanism, the liver proteomes from three groups of rats fed either a normal control diet (NCD), a high fat diet (HFD), or a HFD supplemented with high-dose Fuzhuan brick tea extract (FTE) (HFD + HFTE) were comprehensively compared by quantitative proteomics using 2DE-LC-MS/MS. This is the first study of the effects of tea aqueous extract on the liver proteome of rats suffering from metabolic syndrome. The results showed that 57 proteins displayed more than 1.5-fold differences in at least one of two comparisons of HFD versus NCD and HFD versus HFD + HFTE due to HFD feeding and FTE treatment, respectively. Of them, over 75% of proteins exhibited a similar tendency of expression in the two comparisons, meaning FTE was able to correct HFD effects on rat livers. By function analyses, an extensive list of proteins was involved in sugar and lipid metabolism. Compared with HFD-fed rats, the reduced lipogenesis and enhanced β-oxidation, tricarboxylic acid cycle and respiratory chain in HFD + HFTE-fed rats, which mainly contributed to ameliorate hepatic fat accumulation and associated NAFLD. Additionally, some putative drug targets were also revealed such as COX2, PGAM1, ACACB, FAS, and ECHS1.
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Bromophenol Blue
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)