Merck
  • A disintegrin and metallproteinase 15 knockout decreases migration of fibroblast-like synoviocytes and inflammation in rheumatoid arthritis.
A disintegrin and metallproteinase 15 knockout decreases migration of fibroblast-like synoviocytes and inflammation in rheumatoid arthritis.
Molecular medicine reports (2015-02-05)
Jinliang Gao, Wei Zheng, Liming Wang, Bailin Song
ABSTRACT
The aim of the present study was to determine whether the expression of A disintegrin and metallproteinase 15 (ADAM15) affected the inflammatory conditions and cell migration in human fibroblast‑like synoviocytes (FLSs) in a rat model of rheumatoid arthritis (RA). The expression of ADAM15 in FLSs stimulated with lipopolysaccharide (LPS) was confirmed by reverse transcription‑quantitative polymerase chain reaction and western blot analysis. The effects of small interfering RNA targeting ADAM15 (siADAM5) on pro‑inflammatory cytokines and chemokines were assessed using an enzyme‑linked immunosorbent assay. The effects of siADAM15 on cell invasion and migration in FLS were also assessed in vitro. The therapeutic effects and side effects of ADAM15 in a rat model of collagen‑induced arthritis (CIA) were examined in vivo. The present results revealed that ADAM15 expression was significantly elevated at the mRNA and protein level in FLSs stimulated with LPS and that silencing ADAM15 suppressed the expression of pro‑inflammatory cytokines and chemokines, preventing FLS cell migration and invasion via inhibiting vascular endothelial growth factor‑A, matrix metalloproteinase (MMP)1 and MMP‑3 expression. In addition, treatment of CIA rats using siADAM15 significantly reduced the arthritis score and extent of joint damage in the rats. These findings indicated that silencing ADAM15 had anti‑inflammatory effects in FLSs and efficiently inhibited the development of CIA. Therefore, ADAM15 may be a potential target molecule for RA therapies.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting human MMP3
Sigma-Aldrich
VEGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
MMP-3 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Vascular Endothelial Growth Factor human, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human ADAM15