Merck
  • Combination of cadmium and high cholesterol levels as a risk factor for heart fibrosis.
Combination of cadmium and high cholesterol levels as a risk factor for heart fibrosis.
Toxicological sciences : an official journal of the Society of Toxicology (2015-03-15)
Adrian Türkcan, Bernhard Scharinger, Gerlinde Grabmann, Bernhard K Keppler, Günther Laufer, David Bernhard, Barbara Messner
ABSTRACT
The deleterious effects of increased cadmium (Cd) serum levels on the cardiovascular system are proven by epidemiological and basic science studies. Cd exposure of animals and humans is known to impair myocardial function, possibly leading to heart failure. This study aims at investigating the effect of Cd treatment on the cardiac system with emphasis on the combined effect of Cd and high serum cholesterol levels as an important cardiovascular risk factor. Detailed analyses of Cd-induced effects on the heart of ApoE-/- mice fed a high fat diet (HFD), ApoE-/- mice fed a normal diet (ND), and C57BL/6J mice fed a ND revealed proinflammatory and fibrotic changes in the presence of cellular hypertrophy but in the absence of organ hypertrophy. Hypercholesterolemia in ApoE-/- mice alone and in combination with Cd treatment resulted in significant cardiomyocyte cell death. Based on further analyses of heart sections, we conclude that severe hypercholesterolemia in combination with ApoE-/- genotype as well as Cd treatment results in necrotic cardiomyocyte death. These data were supported by in vitro experiments showing a Cd-induced depolarization of the mitochondrial membrane and the permeabilization of the plasma membrane arguing for the occurrence of Cd-induced necrotic cell death. In summary, we were able to show for the first time that the combination of high cholesterol and Cd levels increase the risk for heart failure through cardiac fibrosis. This observation could in part be explained by the dramatically increased deposition of Cd in the hearts of ApoE-/- mice fed a HFD.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cadmium chloride