  • Search Results
  • Developing a preventive immunization approach against insect bite hypersensitivity using recombinant allergens: A pilot study.
Developing a preventive immunization approach against insect bite hypersensitivity using recombinant allergens: A pilot study.
Veterinary immunology and immunopathology (2015-05-26)
Sigridur Jonsdottir, Eman Hamza, Jozef Janda, Claudio Rhyner, Andreas Meinke, Eliane Marti, Vilhjalmur Svansson, Sigurbjorg Torsteinsdottir
ABSTRACT
Insect bite hypersensitivity (IBH) is an allergic dermatitis of horses caused by bites of midges (Culicoides spp.). IgE-mediated reactions are often involved in the pathogenesis of this disease. IBH does not occur in Iceland due to the absence of Culicoides, but it occurs with a high frequency in Icelandic horses exported to mainland Europe, where Culicoides are present. We hypothesize that immunization with the Culicoides allergens before export could reduce the incidence of IBH in exported Icelandic horses. The aim of the present study was therefore to compare intradermal and intralymphatic vaccination using four purified recombinant allergens, in combination with a Th1 focusing adjuvant. Twelve horses were vaccinated three times with 10μg of each of the four recombinant Culicoides nubeculosus allergens. Six horses were injected intralymphatically, three with and three without IC31(®), and six were injected intradermally, in the presence or absence of IC31(®). Antibody responses were measured by immunoblots and ELISA, potential sensitization in a sulfidoleukotriene release test and an intradermal test, cytokine and FoxP3 expression with real time PCR following in vitro stimulation of PBMC. Immunization with the r-allergens induced a significant increase in levels of r-allergen-specific IgG1, IgG1/3, IgG4/7, IgG5 and IgG(T). Application of the r-allergens in IC31(®) adjuvant resulted in a significantly higher IgG1, IgG1/3, IgG4/7 allergen-specific response. Intralymphatic injection was slightly more efficient than intradermal injection, but the difference did not reach significance. Testing of the blocking activity of the sera from the horses immunized intralymphatically with IC31(®) showed that the generated IgG antibodies were able to partly block binding of serum IgE from an IBH-affected horse to these r-allergens. Furthermore, IgG antibodies bound to protein bands on blots of C. nubeculosus salivary gland extract. No allergen-specific IgE was induced and there was no indication of induction of IgE-mediated reactions, as horses neither responded to Culicoides extract stimulation in a sulfidoleukotriene release test, nor developed a relevant immediate hypersensitivity reaction to the recombinant allergens in skin test. IL-4 expression was significantly higher in horses vaccinated intralymphatically without IC31(®), as compared to horses intradermally vaccinated with IC31(®). Both routes gave higher IL-10 expression with IC31(®). Both intralymphatic and intradermal vaccination of horses with recombinant allergens in IC31(®) adjuvant induced an immune response without adverse effects and without IgE production. The horses were not sensitized and produced IgG that could inhibit allergen-specific IgE binding. We therefore conclude that both the injection routes and the IC31(®) adjuvant are strong candidates for further development of immunoprophylaxis and therapy in horses.
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Diethanolamine, reagent grade, ≥98.0%
Sigma-Aldrich
Diethanolamine, ACS reagent, ≥98.5%
Sigma-Aldrich
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Diethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents