  • Search Results
  • Effect of long-term olanzapine treatment on meal-induced insulin sensitization and on gastrointestinal peptides in female Sprague-Dawley rats.
Effect of long-term olanzapine treatment on meal-induced insulin sensitization and on gastrointestinal peptides in female Sprague-Dawley rats.
Journal of psychopharmacology (Oxford, England) (2015-09-10)
Csaba Hegedűs, Diána Kovács, Rita Kiss, Réka Sári, József Németh, Zoltán Szilvássy, Barna Peitl
ABSTRACT
Meal-induced insulin sensitization (MIS), an endogenous adaptive mechanism is activated post-prandially. Reduced MIS leads to diabetes, but its activation improves insulin sensitivity. MIS is preserved to single olanzapine administration, therefore we aimed to investigate the chronic effect of olanzapine on fasted-state insulin sensitivity and on MIS in female Sprague-Dawley rats. Daily food and water intake, stool and urine production and body weight were determined. The MIS was characterized by a rapid insulin sensitivity test. Fasting hepatic and peripheral insulin sensitivity were determined by a hyperinsulinaemic euglycaemic glucose clamping supplemented with radiotracer technique. Fasted and post-prandial blood samples were obtained for plasma insulin, leptin, ghrelin, amylin, GLP-1, GIP, PYY and PP determination. Adiposity was characterized by weighing intra-abdominal and inguinal fat pads. Olanzapine caused hepatic insulin resistance and a reduced metabolic clearance rate of insulin, but the MIS retained its function. Body weight and adiposity were enhanced, but olanzapine failed to increase food intake. Fasting insulin and leptin were elevated and the post-prandial reduction in ghrelin level was inhibited by olanzapine.The MIS remained functionally intact after long-term olanzapine treatment. Altered insulin, leptin and ghrelin levels indicate olanzapine-induced metabolic derangements. Pharmacological activation of MIS could potentially be exploited to treat or prevent olanzapine-induced insulin resistance.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Olanzapine, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C