  • Search Results
  • Levels of polybrominated diphenyl ethers in indoor dust and human exposure estimates from Makurdi, Nigeria.
Levels of polybrominated diphenyl ethers in indoor dust and human exposure estimates from Makurdi, Nigeria.
Ecotoxicology and environmental safety (2015-06-29)
O I Olukunle, O J Okonkwo, R Sha'ato, G A Wase
ABSTRACT
Information on polybrominated diphenyl ethers (PBDEs) in the indoor environment in developing countries is still relatively scarce. In this study, house (n=10) and office (n=11) dusts samples collected from Makurdi, Benue State Nigeria were extracted and analysed for most abundant PBDEs congeners in the environment. Soxhlet extraction followed by GC-EIMS was employed for the measurement of PBDEs (BDE-47, -100, -99, -154, -153, -183 and -209). The mean concentration of ∑7 PBDEs ranged from 57ngg(-1) to 80ngg(-1) and a median value of 45ngg(-1)and 63ngg(-1) were obtained for house and office dust respectively. The daily exposure and ingestion dose estimates were calculated based on the assumption that 30mg and 60mgday(-1) dust represent the ingestion rate. In addition, the corresponding time spent indoors was assumed to be 87.5% (adult) and 69% (children) in homes and 22% in offices and day care. The average value exposure rate of ∑7PBDEs for children and adults were 2ngday(-1) and 0.84ngday(-1) respectively. The results in the present study, showed higher exposure estimates for both children and adults' in house dust from Nigeria compared to South Africa.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Nonane, anhydrous, ≥99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture