  • Search Results
  • Human alpha-fetal protein immunoassay using fluorescence suppression with fluorescent-bead/antibody conjugate and enzymatic reaction.
Human alpha-fetal protein immunoassay using fluorescence suppression with fluorescent-bead/antibody conjugate and enzymatic reaction.
Biosensors & bioelectronics (2015-04-22)
Junhyoung Ahn, Yong-Beom Shin, JaeJong Lee, Min-Gon Kim
ABSTRACT
The aim of the study was to develop a simple and rapid immunoassay using fluorescent microbeads and enzyme-substrate reactions to measure alpha-fetal protein (AFP) concentrations. We demonstrated the functionality of the fluorescent immunosensor using antibody-conjugated fluorescent latex beads (AB-FLBs) and horseradish peroxidase (HRP) to catalyze a reaction, where the products would precipitate and suppress the fluorescence of AB-FLBs. First, the AB-FLBs were incubated with antigen, biotinylated antibodies (bABs), and streptavidin-HRP (SAv-HRP) to form a sandwich-type immunoreaction. The mixture was then filtered through a membrane to concentrate the beads on a small area. After washing to remove unbound bABs and SAv-HRP, a chromogenic HRP substrate and H2O2 were added to form precipitates on the FLB surface. The suppression of the fluorescence was measured with a fluorescent image analyzer system. Under optimized conditions, AFP could be measured at concentrations as low as 1 pg mL(-1) with a dynamic range up to 100 ng mL(-1).
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
4-Chloro-1-naphthol, tablet
Sigma-Aldrich
4-Chloro-1-naphthol, crystalline
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri