Merck
  • Effect of personalized nutritional counseling in maintenance hemodialysis patients.
Effect of personalized nutritional counseling in maintenance hemodialysis patients.
Hemodialysis international. International Symposium on Home Hemodialysis (2015-01-07)
Cristina Antunes Garagarza, Ana Tentúgal Valente, Telma Sobral Oliveira, Cristina Guerreiro Caetano
ABSTRACT
Monitoring nutritional parameters is an integral part of hemodialysis (HD) patient treatment program. The purpose of this study was to evaluate the impact of the personalized nutritional counseling (PNC) on calcium-phosphorus metabolism, potassium, albumin, protein intake, interdialytic weight gain (IDWG), body composition parameters and fluid overload in HD patients. This was a multicenter longitudinal intervention study with 6 months of follow-up and 731 patients on maintenance HD from 34 dialysis units in Portugal were enrolled. Biochemical and body composition parameters were measured at baseline, 1, 3 and 6 months after the PNC. Patient's mean age was 64.9 (95% confidence interval [CI]: 63.8-66.0) years and mean HD time was 59.8 (95% CI: 55.3-64.3) months. Regarding data comparison collected before PNC vs. 6 months after, we obtained, respectively, the following results: patients with normalized protein catabolic rate (nPCR) ≥ 1 g/kg/day = 66.5% vs. 73.5% (P = 0.002); potassium > 5.5 mEq/L = 52% vs. 35.8% (P < 0.001); phosphorus between 3.5 and 5.5 mg/dL = 43.2% vs. 52.5% (P < 0.001); calcium/phosphorus (Ca/P) ratio ≤ 50 mg/dL = 73.2 % vs. 81.4% (P < 0.001); albumin ≥ 4.0 g/dL = 54.8% vs. 55% (P = 0.808); presence of relative overhydration = 22.4% vs. 25% (P = 0.283); IDWG > 4.5% = 22.3% vs. 18.2% (P = 0.068). PNC resulted in a significant decrease in the prevalence of hyperkalemia, hypophosphatemia and also showed amelioration in Ca/P ratio, nPCR and an increase in P of hyphosphatemic patients. Our study suggests that dietetic intervention contributes to the improvement of important nutritional parameters in patients receiving hemodialysis treatment.
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O