Merck
  • A deletion at ADAMTS9-MAGI1 locus is associated with psoriatic arthritis risk.
A deletion at ADAMTS9-MAGI1 locus is associated with psoriatic arthritis risk.
Annals of the rheumatic diseases (2015-05-21)
Antonio Julià, José Antonio Pinto, Jordi Gratacós, Rubén Queiró, Carlos Ferrándiz, Eduardo Fonseca, Carlos Montilla, Juan Carlos Torre-Alonso, Lluís Puig, José Javier Pérez Venegas, Antonio Fernández Nebro, Emilia Fernández, Santiago Muñoz-Fernández, Esteban Daudén, Carlos González, Daniel Roig, José Luís Sánchez Carazo, Pedro Zarco, Alba Erra, José Luís López Estebaranz, Jesús Rodríguez, David Moreno Ramírez, Pablo de la Cueva, Francisco Vanaclocha, Enrique Herrera, Santos Castañeda, Esteban Rubio, Georgina Salvador, César Díaz-Torné, Ricardo Blanco, Alfredo Willisch Domínguez, José Antonio Mosquera, Paloma Vela, Jesús Tornero, Simón Sánchez-Fernández, Héctor Corominas, Julio Ramírez, María López-Lasanta, Raül Tortosa, Nuria Palau, Arnald Alonso, Andrés C García-Montero, Josep Lluís Gelpí, Laia Codó, Kenneth Day, Devin Absher, Richard M Myers, Juan D Cañete, Sara Marsal
ABSTRACT
Copy number variants (CNVs) have been associated with the risk to develop multiple autoimmune diseases. Our objective was to identify CNVs associated with the risk to develop psoriatic arthritis (PsA) using a genome-wide analysis approach. A total of 835 patients with PsA and 1498 healthy controls were genotyped for CNVs using the Illumina HumanHap610 BeadChip genotyping platform. Genomic CNVs were characterised using CNstream analysis software and analysed for association using the χ(2) test. The most significant genomic CNV associations with PsA risk were independently tested in a validation sample of 1133 patients with PsA and 1831 healthy controls. In order to test for the specificity of the variants with PsA aetiology, we also analysed the association to a cohort of 822 patients with purely cutaneous psoriasis (PsC). A total of 165 common CNVs were identified in the genome-wide analysis. We found a highly significant association of an intergenic deletion between ADAMTS9 and MAGI1 genes on chromosome 3p14.1 (p=0.00014). Using the independent patient and control cohort, we validated the association between ADAMTS9-MAGI1 deletion and PsA risk (p=0.032). Using next-generation sequencing, we characterised the 26 kb associated deletion. Finally, analysing the PsC cohort we found a lower frequency of the deletion compared with the PsA cohort (p=0.0088) and a similar frequency to that of healthy controls (p>0.3). The present genome-wide scan for CNVs associated with PsA risk has identified a new deletion associated with disease risk and which is also differential from PsC risk.
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Supelco
N,N′-Bis(acryloyl)cystamine, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Ammonium hydrogen difluoride, 99.999% trace metals basis
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)