  • Search Results
  • Tanshinone IIA inhibits acute promyelocytic leukemia cell proliferation and induces their apoptosis in vivo.
Tanshinone IIA inhibits acute promyelocytic leukemia cell proliferation and induces their apoptosis in vivo.
Blood cells, molecules & diseases (2015-11-26)
Kaiji Zhang, Jian Li, Wentong Meng, Hongyun Xing, Yiming Yang
ABSTRACT
Tanshinone IIA (TanIIA) is a traditional Chinese agent and has been widely used for treatment of cardiovascular diseases. Our previous study has shown that TanIIA can induce the differentiation of acute promyelocytic leukemia (APL) cells by increasing C/EBPβ expression and induce APL cell apoptosis in vitro. In this study, we evaluated the activity of TanIIA against APL in vivo. We found that treatment with TanIIA prevented APL-mediated reduction in body weights. Treatment with TanIIA inhibited the proliferation of APL cells and triggered APL cell apoptosis and differentiation in vivo. Treatment with TanIIA significantly prolonged the survival of APL-bearing mice. Our data indicate that TanIIA has potent anti-APL activity with little adverse effect.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)