  • Search Results
  • A phosphomimetic mutant TDP-43 (S409/410E) induces Drosha instability and cytotoxicity in Neuro 2A cells.
A phosphomimetic mutant TDP-43 (S409/410E) induces Drosha instability and cytotoxicity in Neuro 2A cells.
Biochemical and biophysical research communications (2015-06-24)
Ki Yoon Kim, Hee-Woo Lee, Yu-Mi Shim, Inhee Mook-Jung, Gye Sun Jeon, Jung-Joon Sung
ABSTRACT
Two DNA/RNA binding proteins, TDP-43 and FUS/TLSU, are involved in RNA processing, and their aberrant mutations induce inherited amyotrophic lateral sclerosis and frontotemporal lobar degeneration with ubiquitinated inclusions. Wild type TDP-43 and FUS (wtTDP-43 and wtFUS) are mainly localized in the nucleus and biochemically interact with the microRNA processing enzyme Drosha. In this study, we investigated Drosha stability in Neuro 2A cells by gain and loss of function studies of wtTDP-43 and wtFUS and cycloheximide mediated protein degradation assay. We also generated three different phosphomimetic mutants of TDP-43 (S379E, S403/404E and S409/410E) by using a site-directed mutagenesis method and examined Drosha stability to elucidate a correlation between the phosphorylated TDP-43 mutants and Drosha stability. Overexpression of wtTDP-43 and/or wtFUS increased Drosha stability in Neuro 2A cells and double knockdown of wtTDP-43 and wtFUS reduced its stability. However, knockdown of wtTDP-43 or wtFUS did not affect Drosha stability in Neuro 2A cells. Interestingly, a phosphomimetic mutant TDP-43 (S409/410E) significantly reduced Drosha stability via prevention of protein-protein interactions between wtFUS and Drosha, and induced cytotoxicity in Neuro 2A cells. Our findings suggest that TDP-43 and FUS controls Drosha stability in Neuro 2A cells and that a phosphomimetic mutant TDP-43 (S409/410E) which is associated with Drosha instability can induce neuronal toxicity.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium fluoride solution
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis