Merck
  • Pro-oxidant HDL predicts poor outcome in patients with ST-elevation acute coronary syndrome.
Pro-oxidant HDL predicts poor outcome in patients with ST-elevation acute coronary syndrome.
Thrombosis and haemostasis (2015-04-03)
Klaus Distelmaier, Lore Schrutka, Veronika Seidl, Max P Winter, Raphael Wurm, Andreas Mangold, Thomas Perkmann, Gerald Maurer, Christopher Adlbrecht, Irene M Lang
ABSTRACT
Oxidative stress affects clinical outcome in patients with ST-elevation acute coronary syndrome (STE-ACS). Although high-density lipoprotein (HDL) particles are generally considered protective, deleterious properties of HDL have been observed in patients with acute myocardial infarction. Here, we analysed the association between pro-oxidant HDL and all-cause mortality in STE-ACS patients. We determined the antioxidant function of HDL in 247 prospectively enrolled patients undergoing primary percutaneous coronary intervention for STE-ACS. Patients were stratified as by a pro-oxidant serum HDL oxidant index (HOI≥ 1) or with an antioxidant serum HOI (HOL< 1) capacity. Multivariate regression analysis was used to relate HOI to survival. The median follow-up time was 23 months (IQR 14.4-40.0 months). Pro-oxidant HDL was observed in 44.1 % of STE-ACS patients and was independently associated with all-cause mortality with a hazard ratio of 3.30(95 %CI 1.50-7.27, p = 0.003). Mortality rates were higher in patients with baseline pro-oxidant HDL compared to patients with preserved HDL function at 30 days (11.9 % vs 2.2 %, p=0.002), and at 4 years (22.9 % vs 8.7 %, p=0.002). Elevated neutrophil counts were a strong and independent predictor for pro-oxidant HDL with an odds ratio per standard deviation of 1.50 (95 %CI 1.11-2.03, p=0.008), as was history of prior acute myocardial infarction, elevated triglycerides levels and reduced glomerular filtration rate. In conclusion, pro-oxidant HDL represents a strong and independent predictor of long-term as well as short-term all-cause mortality in STE-ACS patients. Elevated neutrophil counts predicted the presence of serum pro-oxidant HDL. The maintenance of HDL functions might be a promising therapeutic target in STE-ACS patients.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications