  • Search Results
  • Biocompounds Attenuating the Development of Obesity and Insulin Resistance Produced by a High-fat Sucrose Diet.
Biocompounds Attenuating the Development of Obesity and Insulin Resistance Produced by a High-fat Sucrose Diet.
Natural product communications (2015-10-06)
Usune Etxeberria, Ana Laura de la Garza, J Alfredo Martíinez, I Milagro
ABSTRACT
The use of biocompounds as agents with potential anti-obesity effects might be a feasible alternative to the prescription of traditional drugs in the near future. The goal of the present study was to screen five different compounds in relation to their ability to prevent body weight gain and ameliorate obesity-associated metabolic impairments, namely insulin resistance. For this purpose, seventy Wistar rats were randomly assigned into seven experimental groups. A standard diet-fed control group (control, n=10); a high-fat, high-sucrose diet-fed group (HFS, n=10) and five experimental groups which were fed the HFS diet supplemented with one of the following biocompounds; curcumin (100 mg/kg bw, n=10), chlorogenic acid (50 mg/kg bw, n=10), coumaric acid (100 mg/kg bw, n=10), naringin (100 mg/kg bw, n=10) and leucine (1% of diet, n=10). These results confirm the effectiveness of all the compounds to reduce significantly food efficiency, despite the significant higher food intake. Moreover, visceral fat mass percentage was significantly decreased after naringin and coumaric acid supplementation. In fact, this finding might be related to the considerable amelioration of HOMA-IR index detected in naringin-treated animals. A significant reduction in serum insulin levels and an improvement in the intraperitoneal glucose tolerance test and AUC were found in leucine- and coumaric acid-treated rats, respectively. In summary, the tested biocompounds, particularly naringin, coumaric acid and leucine, showed potential benefits in the prevention of obesity-related complications in rats, at least at the proved doses.
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Naringin, ≥95% (HPLC)
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Naringin, ≥90%, from citrus fruit
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder