  • Search Results
  • Chiral separation of asenapine enantiomers by capillary electrophoresis and characterization of cyclodextrin complexes by NMR spectroscopy, mass spectrometry and molecular modeling.
Chiral separation of asenapine enantiomers by capillary electrophoresis and characterization of cyclodextrin complexes by NMR spectroscopy, mass spectrometry and molecular modeling.
Journal of pharmaceutical and biomedical analysis (2015-10-07)
Zoltán-István Szabó, Gergő Tóth, Gergely Völgyi, Balázs Komjáti, Gabriel Hancu, Lajos Szente, Tamás Sohajda, Szabolcs Béni, Daniela-Lucia Muntean, Béla Noszál
ABSTRACT
The enantiomers of asenapine maleate (ASN), a novel antipsychotic against schizophrenia and mania with bipolar I disorder have been separated by cyclodextrin (CD) modified capillary zone electrophoresis for the first time. 15 different CDs were screened as complexing agents and chiral selectors, investigating the stability of the inclusion complexes and their enantiodiscriminating capacities. Although initially, none of the applied chiral selectors gave baseline separation, β-CD proved to be the most effective chiral selector. In order to improve resolution, an orthogonal experimental design was employed, altering the concentration of background electrolyte, organic modifier, pH, capillary temperature and applied voltage in a multivariate manner. The developed method (160 mM TRIS-acetate buffer pH 3.5, 7 mM β-CD, at 20 °C, applying 15 kV) was successful for baseline separation of ASN enantiomers (R(s)=2.40±0.04). Our method was validated according to ICH guidelines and proved to be sensitive, linear, accurate and precise for the chiral separation of ASN. Properties of the inclusion complexes, such as stoichiometry, atomic level intermolecular host-guest connections are proposed on the basis of ROESY NMR measurement, ESI-MS spectrometry and molecular modeling studies. It was found that the ASN-β-CD complex is of 1:1 composition, and either of the aromatic rings can be accommodated in the β-CD cavity.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
1-Octanol, ACS reagent, ≥99%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Methanol, NMR reference standard