Merck
  • Nutlin-3 loaded nanocarriers: Preparation, characterization and in vitro antineoplastic effect against primary effusion lymphoma.
Nutlin-3 loaded nanocarriers: Preparation, characterization and in vitro antineoplastic effect against primary effusion lymphoma.
International journal of pharmaceutics (2015-05-20)
D Belletti, G Tosi, G Riva, I Lagreca, M Galliania, M Luppi, M A Vandelli, F Forni, B Ruozi
ABSTRACT
In this investigation, Nutlin-3 (Nut3), a novel antitumor drug with low water solubility (<0.1mg/L at 25°C), was loaded into liposomes (Lipo-Nut3), polymeric nanoparticles (NPs-Nut3) and nanoparticles engineered with an antibody direct against Syndecan-1/CD 138 (Syn-NPs-Nut3) to obtain carriers targeted to PEL (primary effusion lymphoma). The physicochemical properties of these carriers were determined. Atomic force microscopy showed that all the particles were well formed and spherical in shape. The presence of the antibody on surface led to a significant increase of mean diameter (280 ± 63 nm), PDI (0.3) and the shift of zeta potential towards neutrality (-1 mV). The entrapment efficiency of Lipo-Nut3, NPs-Nut3 and Syn-NPs-Nut3 was 30, 52 and 29%, and drug loading was 1.4, 4.5 and 2.6%, respectively. By performing cytofluorimetric analyses and bromodeoxyuridine (BrdU) assay, the efficacy of nanocarriers to deliver the antineoplastic drug into a PEL cell line namely BCBL-1 (immortalized body cavity B-cell lymphoma) was investigated. Two days after the treatment with 20 μM of Syn-NPs-Nut3, the cell density decreased at about 60% while the cell viability decreased at 56% only 5 days after transfection, when compared with untreated cells. A cell cycle arrest was observed with a significant decrease of cells in S-phase and increasing of apoptotic cell, if compared with untreated control. These results confirms the potential of nanocarriers approaches to deliver antitumor drug with unfavorable chemico-physical properties. Moreover, this study strongly suggests that Syn-NPs-Nut3 can be a valuable drug carrier system for the treatment of PEL lymphoma.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Nutlin-3, ≥98% (HPLC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%