  • Search Results
  • Characterization and Physicochemical Properties of Condensed Tannins from Acacia catechu.
Characterization and Physicochemical Properties of Condensed Tannins from Acacia catechu.
Journal of agricultural and food chemistry (2016-02-09)
Antoine Duval, Luc Avérous
ABSTRACT
Condensed tannins from Acacia catechu were carefully studied to determine their chemical structure and physicochemical properties. The combined use of MALDI-TOF-MS and (13)C NMR revealed that catechin and epicatechin are the predominant monomers. Most of the compounds were dimers, as confirmed by size exclusion chromatography measurements. To evaluate their potential as aromatic building block in polymer synthesis, special care was given to the characterization and quantification of the different OH groups. A detailed (31)P NMR analysis showed the predominance of catechin, with a catechin/epicatechin ratio of 4.2:1. Two distinct (1)H NMR measurements confirmed the quantification. The thermal properties were also determined: the tannins showed a high temperature of degradation (ca. 190 °C) and a high glass transition temperature (ca. 140 °C), allowing for thermal processing or chemical reactions at relatively high temperature. A. catechu tannins thus present interesting features to be used as aromatic building blocks in polymer materials.
Sigma-Aldrich
2-Chloro-4,4,5,5-tetramethyl-1,3,2-dioxaphospholane, 95%
Sigma-Aldrich
2,3,4,5,6-Pentafluorobenzaldehyde, 98%
Sigma-Aldrich
2-Hydroxy-5-methoxybenzoic acid, 98%