Merck
  • Molecular cloning and characterization of a Glutathione S-transferase gene from Hessian fly (Diptera: Cecidomyiidae)
Molecular cloning and characterization of a Glutathione S-transferase gene from Hessian fly (Diptera: Cecidomyiidae)
Annals of the Entomological Society of America (2004)
Yoshiyama M and Shukle R H