Merck
  • Antiproliferative activity of the ellagic acid-derived gut microbiota isourolithin A and comparison with its urolithin A isomer: the role of cell metabolism.
Antiproliferative activity of the ellagic acid-derived gut microbiota isourolithin A and comparison with its urolithin A isomer: the role of cell metabolism.
European journal of nutrition (2015-12-19)
Antonio González-Sarrías, María Ángeles Núñez-Sánchez, Rocío García-Villalba, Francisco A Tomás-Barberán, Juan Carlos Espín
ABSTRACT
Urolithins, metabolites produced by the gut microbiota from ellagic acid, have been acknowledged with cancer chemopreventive activity. Although urolithin A (Uro-A) has been reported to be the most active one, 10-50 % of humans can also produce the isomer isourolithin A (IsoUro-A). However, no biological activity for IsoUro-A has been reported so far. Herein, we describe for the first time the antiproliferative effect of IsoUro-A, compared to Uro-A, against both human colon cancer (Caco-2) and normal (CCD18-Co) cell lines. Cell proliferation was evaluated by MTT and Trypan blue exclusion assays. Cell cycle was analyzed by flow cytometry and apoptosis measured by the Annexin V/PI method. Finally, urolithins metabolism was analyzed by HPLC-DAD-MS/MS. IsoUro-A inhibited the proliferation of Caco-2 cells in a time- and dose-dependent manner, though it was significantly lower than Uro-A (IC IsoUro-A exerts strong antiproliferative activity, which is reduced by the extensive glucuronidation at 9-position in cancer cells. Further studies are needed to elucidate whether the in vitro structure-activity relationship found for Uro-A and IsoUro-A plays any role in humans.
Sigma-Aldrich
Urolithin A, ≥97% (HPLC)
Pricing and availability is not currently available.