  • Search Results
  • Sulforaphane Prevents Methylmercury-Induced Oxidative Damage and Excitotoxicity Through Activation of the Nrf2-ARE Pathway.
Sulforaphane Prevents Methylmercury-Induced Oxidative Damage and Excitotoxicity Through Activation of the Nrf2-ARE Pathway.
Molecular neurobiology (2016-01-09)
Shu Feng, Zhaofa Xu, Fei Wang, Tianyao Yang, Wei Liu, Yu Deng, Bin Xu
ABSTRACT
Methylmercury (MeHg) is a prominent environmental neurotoxicant, which induces oxidative damage and an indirect excitotoxicity caused by altered glutamate (Glu) metabolism. However, the interaction between oxidative damage and excitotoxicity in MeHg-exposed rats has not been fully recognized. Here, we explored the interaction between oxidative damage and excitotoxicity and evaluated the preventive effects of sulforaphane (SFN) on MeHg-induced neurotoxicity in rat cerebral cortex. Seventy-two rats were randomly assigned to four groups: control group, MeHg-treated groups (4 and 12 μmol/kg), and SFN pretreatment group. After treatment (28 days), the rats were killed and the cerebral cortex was analyzed. Then, Hg, glutathione (GSH), malondialdehyde (MDA), protein sulfhydryl, protein carbonyl, 8-hydroxy-2-deoxyguanosine (8-OHdG), and the levels of reactive oxygen species (ROS) and apoptosis were examined. Glu and glutamine (Gln) levels, glutamine synthetase (GS), phosphate-activated glutaminase (PAG), superoxide dismutase (SOD), glutathione peroxidase (GSH-Px), Na
Sigma-Aldrich
2′,7′-Dichlorofluorescein diacetate, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Ouabain, European Pharmacopoeia (EP) Reference Standard