Merck
  • Molecular characterization of Ambler class A to D β-lactamases, ISAba1, and integrons reveals multidrug-resistant Acinetobacter spp. isolates in northeastern China.
Molecular characterization of Ambler class A to D β-lactamases, ISAba1, and integrons reveals multidrug-resistant Acinetobacter spp. isolates in northeastern China.
Journal of chemotherapy (Florence, Italy) (2016-04-15)
Xiaoyu Sun, Bin Liu, Yan Chen, Honglan Huang, Guoqing Wang, Fan Li, Zhaohui Ni
ABSTRACT
The prevalence of various Ambler class A to D β-lactamases, ISAba1, and class 1 and 2 integrons as well as the clonal relatedness in 105 Acinetobacter spp. isolates found in northeastern China was investigated. All 105 Acinetobacter spp. isolates were determined to be multidrug resistant (MDR), and the resistance rates to carbapenem agents were approximately 50%. PER, IMP, AmpC, and OXA-23 were found to be dominant β-lactamases belonging to different classes, respectively. This is the first report of the coexistence of bla
Product Description
Millipore
Ticarcillin Supplement, suitable for microbiology