Merck
  • Investigation of mechanisms involved in postprandial glycemia and insulinemia attenuation with dietary fibre consumption.
Investigation of mechanisms involved in postprandial glycemia and insulinemia attenuation with dietary fibre consumption.
Food & function (2017-06-06)
Nikolay Repin, Brittney A Kay, Steve W Cui, Amanda J Wright, Alison M Duncan, H Douglas Goff
ABSTRACT
This work examines the mechanisms involved in the attenuation of postprandial glycemic and insulinemic responses associated with soluble dietary fibre (SDF) consumption. The effect of SDF, including yellow mustard mucilage, soluble flaxseed gum and fenugreek gum on in vitro amylolysis and maltose transport was studied. Furthermore, a human clinical trial was conducted to investigate the effect of SDF consumption on postprandial glycemic and insulinemic responses and gastric emptying, as estimated based on the absorption of paracetamol. Participants (n = 15) at risk for type II diabetes consumed maltose syrup- and starch-based pudding treatments supplemented with each SDF, each at a concentration to match three times the apparent viscosity (18.54 mPa s at 60 s
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology