  • Search Results
  • A novel autophagy modulator 6-Bio ameliorates SNCA/α-synuclein toxicity.
A novel autophagy modulator 6-Bio ameliorates SNCA/α-synuclein toxicity.
Autophagy (2017-03-30)
S N Suresh, Aravinda K Chavalmane, Vidyadhara Dj, Haorei Yarreiphang, Shashank Rai, Abhik Paul, James P Clement, Phalguni Anand Alladi, Ravi Manjithaya
ABSTRACT
Parkinson disease (PD) is a life-threatening neurodegenerative movement disorder with unmet therapeutic intervention. We have identified a small molecule autophagy modulator, 6-Bio that shows clearance of toxic SNCA/α-synuclein (a protein implicated in synucleopathies) aggregates in yeast and mammalian cell lines. 6-Bio induces autophagy and dramatically enhances autolysosome formation resulting in SNCA degradation. Importantly, neuroprotective function of 6-Bio as envisaged by immunohistology and behavior analyses in a preclinical model of PD where it induces autophagy in dopaminergic (DAergic) neurons of mice midbrain to clear toxic protein aggregates suggesting that it could be a potential therapeutic candidate for protein conformational disorders.
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
SAFC
Trypsin-EDTA Solution 10X
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 70,000, (FITC:Glucose = 1:250)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
3,3′-Diaminobenzidine (DAB) Enhanced Liquid Substrate System tetrahydrochloride, for Immunohistology
Sigma-Aldrich
Anti-Amyloid Oligomer Antibody, αβ, oligomeric, serum, Chemicon®
Sigma-Aldrich
Anti-Rabbit IgG - Atto 633 antibody produced in goat, 1 mg/mL IgG