Merck
Cadmium induces Ca
Journal of inorganic biochemistry (2017-04-19)
Hong Wang, Nianhui Zhai, Ying Chen, Haibin Xu, Kehe Huang
ABSTRACT
Calcium, as a ubiquitous second messenger, governs a large array of cellular processes and is necessary for cell survival. More recently, it was observed that the cytosolic Ca
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-al, ≥95%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
PD 150606, ≥97% (HPLC)
Pricing and availability is not currently available.