Merck
  • Matrine prevents bone loss in ovariectomized mice by inhibiting RANKL-induced osteoclastogenesis.
Matrine prevents bone loss in ovariectomized mice by inhibiting RANKL-induced osteoclastogenesis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-07-26)
Xiao Chen, Xin Zhi, Panpan Pan, Jin Cui, Liehu Cao, Weizong Weng, Qirong Zhou, Lin Wang, Xiao Zhai, Qingiie Zhao, Honggang Hu, Biaotong Huang, Jiacan Su
ABSTRACT
Osteoporosis is a metabolic bone disease characterized by decreased bone density and strength due to excessive loss of bone protein and mineral content. The imbalance between osteogenesis by osteoblasts and osteoclastogenesis by osteoclasts contributes to the pathogenesis of postmenopausal osteoporosis. Estrogen withdrawal leads to increased levels of proinflammatory cytokines. Overactivated osteoclasts by inflammation play a vital role in the imbalance. Matrine is an alkaloid found in plants from the
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)