  • Search Results
  • Proton Channel Activity of Influenza A Virus Matrix Protein 2 Contributes to Autophagy Arrest.
Proton Channel Activity of Influenza A Virus Matrix Protein 2 Contributes to Autophagy Arrest.
Journal of virology (2015-10-16)
Yizhong Ren, Chufang Li, Liqiang Feng, Weiqi Pan, Liang Li, Qian Wang, Jiashun Li, Na Li, Ling Han, Xuehua Zheng, Xuefeng Niu, Caijun Sun, Ling Chen
ABSTRACT
Influenza A virus infection can arrest autophagy, as evidenced by autophagosome accumulation in infected cells. Here, we report that this autophagosome accumulation can be inhibited by amantadine, an antiviral proton channel inhibitor, in amantadine-sensitive virus infected cells or cells expressing influenza A virus matrix protein 2 (M2). Thus, M2 proton channel activity plays a role in blocking the fusion of autophagosomes with lysosomes, which might be a key mechanism for arresting autophagy.
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-p62/SQSTM1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution