Merck
  • Two new alpha1-globin gene point mutations: Hb Nedlands (HBA1:c.86C>T) [alpha28(B9)Ala-->Val] and Hb Queens Park (HBA1:c.98T>A) [alpha32(B13)Met-->Lys].
Two new alpha1-globin gene point mutations: Hb Nedlands (HBA1:c.86C>T) [alpha28(B9)Ala-->Val] and Hb Queens Park (HBA1:c.98T>A) [alpha32(B13)Met-->Lys].
Hemoglobin (2010-04-01)
Marion Phylipsen, John F Prior, Erna Lim, Neela Lingam, Jill Finlayson, Sandra G J Arkesteijn, Cornelis L Harteveld, Piero C Giordano
ABSTRACT
We report two new point mutations of the alpha1-globin gene found in a Greek and a Burmese patient, both living in Western Australia. The patients were initially selected for their microcytic hypochromic parameters as belonging to a group suspected for uncommon (deletion) defects. Gap-polymerase chain reaction (gap-PCR) and multiplex ligation-dependent probe amplification (MLPA) technologies were applied, and in those cases not showing deletions, direct sequencing was performed. We have found 1) HBA1:c.86C>T, Hb Nedlands [alpha28(B9)Ala-->Val] which, based on the red cell indices and phenotype prediction scores, is presumed to be clinically silent, and 2) HBA1:c.98T>A, Hb Queens Park [alpha32(B13)Met-->Lys] which seems to be associated with a mild alpha-thalassemia (alpha-thal) phenotype. The phenotype/genotype correlation is briefly described.