Merck
  • Studies on the metabolism and toxicological detection of the new psychoactive designer drug 2-(4-iodo-2,5-dimethoxyphenyl)-N-[(2-methoxyphenyl)methyl]ethanamine (25I-NBOMe) in human and rat urine using GC-MS, LC-MS(n), and LC-HR-MS/MS.
Studies on the metabolism and toxicological detection of the new psychoactive designer drug 2-(4-iodo-2,5-dimethoxyphenyl)-N-[(2-methoxyphenyl)methyl]ethanamine (25I-NBOMe) in human and rat urine using GC-MS, LC-MS(n), and LC-HR-MS/MS.
Analytical and bioanalytical chemistry (2015-06-26)
Achim T Caspar, Andreas G Helfer, Julian A Michely, Volker Auwärter, Simon D Brandt, Markus R Meyer, Hans H Maurer
ABSTRACT
25I-NBOMe, a new psychoactive substance, is a potent 5-HT2A receptor agonist with strong hallucinogenic potential. Recently, it was involved in several fatal and non-fatal intoxication cases. The aim of the present work was to study its phase I and II metabolism and its detectability in urine screening approaches. After application of 25I-NBOMe to male Wistar rats, urine was collected over 24 h. The phase I and II metabolites were identified by LC-HR-MS/MS in urine after suitable workup. For the detectability studies, standard urine screening approaches (SUSA) by GC-MS, LC-MS(n), and LC-HR-MS/MS were applied to rat and also to authentic human urine samples submitted for toxicological analysis. Finally, an initial CYP activity screening was performed to identify CYP isoenzymes involved in the major metabolic steps. 25I-NBOMe was mainly metabolized by O-demethylation, O,O-bis-demethylation, hydroxylation, and combinations of these reactions as well as by glucuronidation and sulfation of the main phase I metabolites. All in all, 68 metabolites could be identified. Intake of 25I-NBOMe was detectable mainly via its metabolites by both LC-MS approaches, but not by the GC-MS SUSA. Initial CYP activity screening revealed the involvement of CYP1A2 and CYP3A4 in hydroxylation and CYP2C9 and CYP2C19 in O-demethylation. The presented study demonstrated that 25I-NBOMe was extensively metabolized and could be detected only by the LC-MS screening approaches. Since CYP2C9 and CYP3A4 are involved in initial metabolic steps, drug-drug interactions might occur in certain constellations.
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Ultrapure Acetonitrile
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles