Skip to Content
Merck

BDPH9BCCCEB2

Bide Pharmatech

4-Chloro-6-methyl-2-(trifluoromethyl)quinoline

Synonym(s):

4-chloro-6-methyl-2-(trifluoromethyl)quinoline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H7N1Cl1F3
CAS Number:
Molecular Weight:
245.63
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H7ClF3N/c1-6-2-3-9-7(4-6)8(12)5-10(16-9)11(13,14)15/h2-5H,1H3

Inchi Key

ZLSXNETUGWRDSH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library