Merck
All Photos(1)

1.06544

SAFC

Sodium nitrite

extra pure Ph Eur,USP

Synonym(s):
Sodium nitrite
Empirical Formula (Hill Notation):
NNaO2
CAS Number:
Molecular Weight:
69.00
MDL number:
EC Index Number:
231-555-9

Agency

Ph. Eur.
USP

Quality Level

form

solid

potency

85 mg/kg LD50, oral (Rat)

pH

9 (20 °C, 100 g/L in H2O)

mp

280 °C (decomposition)

solubility

820 g/L

density

2.1 g/cm3 at 20 °C

bulk density

1200 kg/m3

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sodium nitrite is used as reagent.

Packaging

1, 5 kg in PE bottle
50 kg in PE bag (in carton)

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service