Select a Size
About This Item
Product Name
Anti CXCL5 Antibody, clone 23GB5670, Rabbit Monoclonal, clone 23GB5670, recombinant rabbit monoclonal, expressed in HEK293 cells
biological source
rabbit (Monoclonal)
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
23GB5670, recombinant monoclonal
form
buffered aqueous glycerol solution
mol wt
12 kDa
species reactivity
human
packaging
vial of 1.5 mL
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
Quality Level
Gene Information
human ... CXCL5(6374)
sustainability
Greener Alternative Product
concentration
2 mg/ml
technique(s)
western blot: 1:1,000-1:5,000
isotype
IgG
Ensembl | human accession no.
NCBI accession no.
UniProt accession no.
greener alternative category
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Related Categories
General description
Immunogen
Other Notes
Physical form
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service