Product Name
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride purum, ≥98.0% (AT)
Product Number
03450
Product Brand
sial
CAS Number
25952-53-8
Molecular Weight
191.70
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
ARGENTOMETR. TITRATION98.0 - 102.0 %
MELTING POINT110 - 115 C
SOLUBILITY (COLOR)Colorless to Very Faint Yellow
SOLUBILITY (TURBIDITY)Clear to Very Slightly Hazy
SOLUBILITY (METHOD)1G IN 10ML WATER
WATER≤ 1.5 %
CARBON CONTENT50.12 % (THEORY)
HYDROGEN CONTENT9.46 % (THEORY)
NITROGEN CONTENT21.92 % (THEORY)
INFRARED SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD36 MONTHS