Product Name
(±)-Sulpiride
Product Number
s8010
Product Brand
sigma
CAS Number
15676-16-1
Molecular Weight
341.43
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder to Crystals
TITRATION (NT) HCLO4 0.1M≥ 98%
PURITY (TLC AREA %)≥ 98 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)50 mg/ml in Acetic acid
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD72 MONTHS