Skip to Content
Merck
Ammonium thiocyanate solution c(NH4SCN) = 0.1 mol/l (0.1 N), Titripur®, reag. Ph. Eur., reag. USP

Ammonium thiocyanate solution

Ammonium thiocyanate solution c(NH4SCN) = 0.1 mol/l (0.1 N), Titripur®, reag. Ph. Eur., reag. USP
Synonym(s):
Ammoniumrhodanid solution

Products

Product Category
Brand
Physical Form
Product Line
Quality Segment
Technique