Skip to Content
Merck
4-methylhexan-1-ol

4-methylhexan-1-ol

4-methylhexan-1-ol
Synonym(s):
4-methyl-1-hexanol
Linear Formula:
C7H16O1
CAS Number:
Molecular Weight:
116.2

Products

Brand
Physical Form
Showing 1-2 of 2
Compare
Product Number
Description
Pricing