Skip to Content
Products
Cart
0
CZ
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Caco-2 Engineered Control Cells
Synonym(s):
Caco-2 cell line
Products
Product Category
cell lines
Brand
Sigma-Aldrich
Biological Source
Physical Form
Tissue
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
MTOX1000CC
Caco-2 Engineered Control Cells,
one vial of frozen cells
Expand
View Pricing
An unknown error has occured.