Skip to Content
Merck
Ephos ≥95%

Ephos

Ephos ≥95%
Synonym(s):
Dicyclohexyl(3-isopropoxy-2′,4′,6′-triisopropyl-[1,1′-biphenyl]-2-yl)phosphane
Empirical Formula (Hill Notation):
C36H55OP
CAS Number:
Molecular Weight:
534.80

Products

Product Category
Brand
Sustainability
Physical Form
Product Line
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing