Skip to Content
Products
Cart
0
CZ
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Ferroin solution
Synonym(s):
1,10-Phenanthroline iron(II) salt solution
Products
Product Category
indicators
reagents
Brand
Supelco
Physical Form
Quality Segment
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1.09193
Ferroin solution,
(1,10-phenanthroline iron(II) sulfate) 1/40 mol/l redox indicator
Expand
View Pricing
An unknown error has occured.