
Gram’s crystal violet solution

Synonym(s):
Basic violet 3; gentian violet; hexamethyl pararosaniline; methyl violet 10B
Products
Product Category
Brand
Color
Physical Form
Product Line
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing