
MILLIPLEX® Humanized Mouse Panel

Synonym(s):
Luminex® humanized mouse assay, Millipore humanized mouse panel, humanized mouse multiplex kit
Products
Product Category
Product Line
Detection Method
Quality Segment
Species Reactivity
Technique
Showing 1-2 of 2