Skip to Content
Merck
Normal Rabbit IgG Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.

Normal Rabbit IgG

Normal Rabbit IgG Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Synonym(s):
Rabbit Immunoglobulin

Products

Product Category
Brand
Biological Source
Clonality
Physical Form
Conjugate
Quality Segment
Technique