Skip to Content
Merck
vibrant-m

Oxycodone impurity D

vibrant-m
Synonym(s):
14-Hydroxycodeinone, (5α)-7,8-Didehydro-4,5-epoxy-14-hydroxy-3-methoxy-17-methylmorphinan-6-one
Empirical Formula (Hill Notation):
C18H19NO4
CAS Number:
Molecular Weight:
313.35

Products

Product Category
Manufacturer
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing