Skip to Content
Merck
Poly(A) lyophilized, suitable for PCR, pkg of 100 mg

Poly(A)

Poly(A) lyophilized, suitable for PCR, pkg of 100 mg
Synonym(s):
Poly(A), Polyadenylic acid

Products

Product Category
Color
Manufacturer
Physical Form
Quality Segment
Technique